Learn More
Metrohm For use with Metrohm KF Titrators.
Product Code. 10391245
Encompass_Preferred
Change view
Click to view available options
Type:
Titration Vessel (90 mL)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type unitSize
10391245 Titration Vessel (90 mL) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10391245 Supplier Metrohm Supplier No. 61415220
Only null left
Add to basket
Add to basket
  • Accessories and replacement parts
Specifications
Diameter (English) 3.07 in.
Diameter (Metric) 78 mm
Type Titration Vessel (90 mL)
Height (English) 3.14 in.
Height (Metric) 80 mm
Material Clear Glass
Volume (Metric) 20 to 90 mL