Learn More
Metrohm Electrolyte Solutions for Metrohm™ Karl Fischer Titrator Electrodes
Product Code. 12063491
Encompass
Change view
Click to view available options
Filling Solution Type:
Potassium Chloride Electrolyte
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Filling Solution Type Unit Size
12063491 Potassium Chloride Electrolyte Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12063491 Supplier Metrohm Supplier No. 62308020
Estimated Shipment: 25-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
For Use With (Equipment) Metrohm™ Karl Fischer Titrator Electrode, Ag/AgCl Reference Systems
Type Electrolyte Solution
Filling Solution Type Potassium Chloride Electrolyte
Volume (Metric) Sample 250 mL