missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brivanib Alaninate, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Brivanib Alaninate |
| CAS | 649735-63-7 |
| Molecular Formula | C22 H24 F N5 O4 |
| Quantity | 10 mg |
| Synonym | L-Alanine (1R)-2-[[4-[(4-fluoro-2-methyl-1H-indol-5-yl)oxy]-5-methylpyrrolo [2,1-f][1,2,4]triazin-6-yl]oxy]-1-methylethyl Ester, BMS 582664 |
| InChI Formula | InChI=1S/C22H24FN5O4/c1-11-7-15-16(27-11)5-6-17(19(15)23)32-21-20-13(3)18(8-28(20)26-10-25-21)30-9-12(2)31-22(29)14(4)24/h5-8,10,12,14,27H,9,24H2,1-4H3/t12-,14+/m1/s1 |
| SMILES | C[C@H](COc1cn2ncnc(Oc3ccc4[nH]c(C)cc4c3F)c2c1C)OC(=O)[C@H](C)N |
| IUPAC Name | [(2R)-1-[4-[(4-fluoro-2-methyl-1H-indol-5-yl)oxy]-5-methylpyrrolo[2,1-f][1,2,4]triazin-6-yl]oxypropan-2-yl] (2S)-2-aminopropanoate |
| Molecular Weight (g/mol) | 441.455 |
| Formula Weight | 441.181 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction