missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRIX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15624
This item is not returnable.
View return policy
Description
BRIX Polyclonal specifically detects BRIX in Human samples. It is validated for Western Blot.
Specifications
| BRIX | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| brix domain containing 2, Brix domain-containing protein 2, BRIXBXDC2, BRX1, biogenesis of ribosomes, homolog (S. cerevisiae), FLJ11100, ribosome biogenesis protein BRX1 homolog | |
| Rabbit | |
| 41 kDa | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 55299 | |
| Human | |
| IgG |
| Western Blot | |
| 0.79mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q8TDN6 | |
| BRIX1 | |
| Recombinant protein encompassing a sequence within the center region of human BXDC2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Mouse (97%), Xenopus laevis (82%), Bovine (99%), Xenopus tropicalis (81%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction