missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRIX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99257-100ul
This item is not returnable.
View return policy
Description
BRIX Polyclonal antibody specifically detects BRIX in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| BRIX | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:300-1:10000 | |
| brix domain containing 2, Brix domain-containing protein 2, BRIXBXDC2, BRX1, biogenesis of ribosomes, homolog (S. cerevisiae), FLJ11100, ribosome biogenesis protein BRX1 homolog | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human BRIX. | |
| 100 μg | |
| Core ESC Like Genes, Stem Cell Markers | |
| 55299 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction