missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brk Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98347AF350
This item is not returnable.
View return policy
Description
Brk Polyclonal specifically detects Brk in Mouse samples. It is validated for ELISA.
Specifications
| Brk | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| ELISA | |
| Alexa Fluor 350 | |
| Breast tumor kinase, BRKprotein-tyrosine kinase 6, EC 2.7.10, EC 2.7.10.2, FLJ42088, protein-tyrosine kinase BRK, PTK6 protein tyrosine kinase 6, Tyrosine-protein kinase BRK | |
| Produced in rabbits immunized with purified, recombinant Brk (Q64434; Met1-Val451). | |
| 0.1 mL | |
| Protein Kinase | |
| 5753 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction