Learn More
Invitrogen™ BRK Monoclonal Antibody (2H12B8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515328
Description
MA5-15328 targets PTK6 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15328 immunogen is purified recombinant fragment of human PTK6 expressed in E. Coli. . MA5-15328 detects PTK6 which has a predicted molecular weight of approximately 52kDa.
Breast tumor kinase (Brk) is a non-receptor protein tyrosine kinase expressed in over 60% of breast carcinoma tissue samples and breast tumor cell lines. Brk is 80% identity to Sik (Src-related intestinal kinase) and exhibits the features of a novel non-receptor tyrosine kinase, including amino terminal SH3 and SH2 domains. Low levels of Brk transcripts have been identified in some human breast tumor cell lines but not in normal breast tissue.
Specifications
| BRK | |
| Monoclonal | |
| Conc. Not Determined | |
| ascites with 0.03% sodium azide | |
| Q13882 | |
| PTK6 | |
| Purified recombinant fragment of human PTK6 expressed in E. Coli. | |
| RUO | |
| 5753 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 2H12B8 | |
| Unconjugated | |
| PTK6 | |
| ALT-PTK6; breast tumor kinase; BRK; deltam5; FLJ42088; protein tyrosine kinase 6; protein-tyrosine kinase 6; protein-tyrosine kinase BRK; Ptk6; PTK6 protein tyrosine kinase 6; Sik; SRC-related intestinal kinase; tks; Tksk; tyrosine kinase identified from skin; tyrosine-protein kinase BRK | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.