missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRMS1 Antibody (SR1460), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22436-50ul
This item is not returnable.
View return policy
Description
BRMS1 Monoclonal antibody specifically detects BRMS1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Specifications
| BRMS1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| SR1460 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
| breast cancer metastasis suppressor 1, breast cancer metastasis-suppressor 1, DKFZP564A063 | |
| A synthesized peptide derived from human BRMS1 (Uniprot #: Q9HCU9) | |
| 50 μg | |
| Cell Cycle and Replication | |
| 25855 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction