Learn More
Invitrogen™ Brodalumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558330
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Brodalumab has been used in trials studying the treatment of Asthma, Psoriasis, Crohn's Disease, Psoriatic Arthritis, and Rheumatoid Arthritis. Brodalumab was FDA approved in February, 2017 as Siliq for the treatment of moderate-to-severe plaque psoriasis.
Specifications
| Brodalumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| AMG827; KHK4827 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG2SA |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 1 mg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |