Learn More
Bromacil, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | Bromacil |
| CAS | 314-40-9 |
| Molecular Formula | C9 H13 Br N2 O2 |
| Quantity | 25 g |
| Synonym | Uracil, 5-bromo-3-sec-butyl-6-methyl- (8CI), 5-Bromo-6-methyl-3-(1-methylpropyl)-2,4(1H,3H)-pyrimidinedione, (±)-Bromacil, 3-sec-Butyl-5-bromo-6-methyluracil, 5-Bromo-3-sec-butyl-6-methyluracil, 5-Bromo-6-methyl-3-(1-methyl-n-propyl)uracil, 5-Bromo-6-methyl-3-(1-methylpropyl)uracil, 5-Bromo-6-methyl-3-sec-butyluracil, Bromacil, DuPont 976, Herbicide 976, Hyvar X, Hyvar X bromacil, Hyvarex, Uragan |
| InChI Formula | InChI=1S/C9H13BrN2O2/c1-4-5(2)12-8(13)7(10)6(3)11-9(12)14/h5H,4H2,1-3H3,(H,11,14) |
| SMILES | CCC(C)N1C(=O)NC(=C(Br)C1=O)C |
| IUPAC Name | 5-bromo-3-butan-2-yl-6-methyl-1H-pyrimidine-2,4-dione |
| Molecular Weight (g/mol) | 261.12 |
| Formula Weight | 260.016 |