missing translation for 'onlineSavingsMsg'
Learn More

Bromfenac sodium hydrate, MedChemExpress

Product Code. 16418728
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
This item is not returnable. View return policy

Product Code. 16418728

Brand: Medchem Express HYB1888B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Bromfenac sodium hydrate (Bromfenac monosodium salt sesquihydrate) is a potent and orally active inhibitor of COX, with IC50s of 5.56 and 7.45 nM for COX-1 and COX-2, respectively. Bromfenac sodium hydrate can be used in ocular inflammation research.

Bromfenac sodium hydrate (Bromfenac monosodium salt sesquihydrate) is a potent and orally active inhibitor of COX, with IC50s of 5.56 and 7.45 nM for COX-1 and COX-2, respectively. Bromfenac sodium hydrate can be used in ocular inflammation research.

Specifications

Chemical Name or Material Bromfenac sodium hydrate
CAS 120638-55-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H14BrNNaO4.5
Quantity 10 mg
Synonym Bromfenac monosodium salt sesquihydrate
Solubility Information DMSO : ≥ 100 mg/mL (260.98 mM) ∣H2O : ≥ 100 mg/mL (260.98 mM)
SMILES O=C(O[Na])CC1=CC=CC(C(C2=CC=C(Br)C=C2)=O)=C1N.[1.5H2O]
Molecular Weight (g/mol) 383.17
Formula Weight 383.17
Percent Purity 99.7%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.