Learn More
Bromfenac Sodium Sesquihydrate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Bromfenac Sodium Sesquihydrate
Specifications
Specifications
| Chemical Name or Material | Bromfenac Sodium Sesquihydrate |
| CAS | 120638-55-3 |
| Analyte or Component Names | Bromfenac Sodium Sesquihydrate |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2 C15 H11 Br N O3 . 2 Na . 3 H2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Bromfenac Sodium Sesquihydrate, Benzeneacetic acid, 2-amino-3-(4-bromobenzoyl)-, sodium salt, hydrate (2:2:3), Benzeneacetic acid, 2-amino-3-(4-bromobenzoyl)-, monosodium salt, hydrate (2:3) (9CI) |
| InChI Formula | InChI=1S/2C15H12BrNO3.2Na.3H2O/c2*16-11-6-4-9(5-7-11)15(20)12-3-1-2-10(14(12)17)8-13(18)19;;;;;/h2*1-7H,8,17H2,(H,18,19);;;3*1H2/q;;2*+1;;;/p-2 |
| SMILES | O.O.O.[Na+].[Na+].Nc1c(CC(=O)[O-])cccc1C(=O)c2ccc(Br)cc2.Nc3c(CC(=O)[O-])cccc3C(=O)c4ccc(Br)cc4 |
| IUPAC Name | disodium;2-[2-amino-3-(4-bromobenzoyl)phenyl]acetate;trihydrate |
| Show More |