Learn More
Bromocresol Purple 0.04% Indicator Solution According to European Pharmacopoeia (EP), Reagecon
BIOPHARMA
Product Code. 11404679 Shop All Reagecon Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Product Code. 11404679
Brand: Reagecon 1012701
High-quality indicator solution produced from the highest quality raw materials and manufactured and tested under rigorous conditions.
  • European Pharmacopoeia (EP) Reference Standard, European Pharmacopoeia (EP) Reference Material
  • High quality indicator solution produced from the highest quality raw materials and manufactured and tested under rigorous conditions.
  • For manual titrations; indicator titrant or analyte change color and this color change is by far the most important method of end point detection, in such titrations.
  • A high-quality indicator solution such as Reagecon's facilitates end point detection, in a clear and unambiguous manner and is an imperative for accurate manual titrimetry.
Specifications
Concentration 0.0004
Packaging HDPE Bottle
Quantity 100 mL
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • BROMOCRESOL PURPLE SOLUTION
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501-Dispose of contents/container to .
Supplemental information
  • MIXTURE LIST-Contains : ethanol