missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromodeoxyuridine/BrdU Antibody (BRD.3), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-92989AF700
This item is not returnable.
View return policy
Description
Bromodeoxyuridine/BrdU Monoclonal specifically detects Bromodeoxyuridine/BrdU in All Species samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| Bromodeoxyuridine/BrdU | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Bromodeoxyuridine, BUdr | |
| Bromodeoxyuridine (BrdU) conjugated to BSA | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| BRD.3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| All species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction