missing translation for 'onlineSavingsMsg'
Learn More

Bromodeoxyuridine/BrdU Antibody (BRD469), Novus Biologicals™

Mouse Monoclonal Antibody

€ 264.00 - € 474.00

Specifications

Antigen Bromodeoxyuridine/BrdU
Clone BRD469
Concentration 0.2 mg/mL
Dilution Immunohistochemistry -frozen 0.5-1.0ug/mL, Immunocytochemistry/ Immunofluorescence 0.5-1ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL
Applications Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18457401
View Documents
Novus Biologicals
NBP2-32918-0.1mg
0.1 mg
€ 474.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18423192
View Documents
Novus Biologicals
NBP2-32918-0.02mg
0.02 mg
€ 264.00
0.02mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Bromodeoxyuridine/BrdU Monoclonal antibody specifically detects Bromodeoxyuridine/BrdU in Nonspecies specific samples. It is validated for Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

Bromodeoxyuridine/BrdU
0.2 mg/mL
Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Neuronal Cell Markers, Neuroscience, Neurotransmission
10 mM PBS with 0.05% BSA
Bromodeoxyuridine/BrdU conjugated to KLH
Primary
Store at 4C.
BRD469
Immunohistochemistry -frozen 0.5-1.0ug/mL, Immunocytochemistry/ Immunofluorescence 0.5-1ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL
Monoclonal
Purified
RUO
Non-species specific
Bromodeoxyuridine, BUdr
IgG1 κ
Protein A or G purified
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.