missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromodeoxyuridine/BrdU Antibody (BU-1), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB7225MFV450
This item is not returnable.
View return policy
Description
Bromodeoxyuridine/BrdU Monoclonal antibody specifically detects Bromodeoxyuridine/BrdU in Broad Species Reactivity samples. It is validated for Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF
Specifications
| Bromodeoxyuridine/BrdU | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. |
| Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF | |
| BU-1 | |
| Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
| Bromodeoxyuridine, BUdr | |
| 5-iodourindine (5-IO) coupled to ovalbumin | |
| RUO | |
| Broad Species Reactivity | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction