missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromodeoxyuridine/BrdU Antibody (rBRD.3), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08847AF405
This item is not returnable.
View return policy
Description
Bromodeoxyuridine/BrdU Monoclonal antibody specifically detects Bromodeoxyuridine/BrdU in Nonspecies specific samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Bromodeoxyuridine/BrdU | |
| Monoclonal | |
| Alexa Fluor 405 | |
| Bromodeoxyuridine, BUdr | |
| Bromodeoxyuridine/BrdU conjugated to BSA | |
| 100 μg | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| Non-species specific | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| rBRD.3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction