Learn More
Bromoethane, 98%
Product Code. 11425173
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Unit Size:
100g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11425173 2500 g 2500g
11404190 100 g 100g
11415173 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product contains an ozone depleters according to Regulation (EC) No 1005/2009, all customers must complete a declaration before delivery.
Product Code. 11425173 Supplier Thermo Scientific Alfa Aesar Supplier No. A12804.0E
  • ONLINE EXCLUSIVE PRICE
    € 108.00
    € 97.65 / 2500g
    Save € 10.35
    9% Off
 Restricted Item
In Stock
Add to basket
Regulatory: This product contains an ozone depleters according to Regulation (EC) No 1005/2009, all customers must complete a declaration before delivery.
CAS: 74-96-4 | C2H5Br | 108.966 g/mol
Bromoethane is used in organic synthesis for the introduction of ethyl group. It is widely used as solvent, as an anesthetic in medicine and as a refrigerant. It is an important component of antiknock fluid in gasoline. It is also also used in the manufacture of pharmaceuticals like vitamin A as well as in the manufacture of flame retardants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bromoethane is used in organic synthesis for the introduction of ethyl group. It is widely used as solvent, as an anesthetic in medicine and as a refrigerant. It is an important component of antiknock fluid in gasoline. It is also also used in the manufacture of pharmaceuticals like vitamin A as well as in the manufacture of flame retardants.
Solubility
Miscible with ethanol, ether, chloroform and organic solvents.
Notes
Light sensitive. Store in cool place. Incompatible with strong bases, strong oxidizing agents, magnesium, potassium, calcium, sodium oxides, aluminum, zinc and alkali metals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 74-96-4
Molecular Formula C2H5Br
Molecular Weight (g/mol) 108.966
MDL Number MFCD00000232
InChI Key RDHPKYGYEGBMSE-UHFFFAOYSA-N
Synonym ethyl bromide, ethane, bromo, 1-bromoethane, monobromoethane, bromic ether, hydrobromic ether, bromure d'ethyle, ethylbromide, etylu bromek, halon 2001
PubChem CID 6332
IUPAC Name bromoethane
SMILES CCBr
Specifications
Melting Point -119°C
Density 1.461
Boiling Point 37°C to 39°C
Flash Point −23°C (−9°F)
Assay Percent Range ≥98%
Odor Ether-like
Linear Formula BrCH2CH3
Refractive Index 1.4235
Quantity 2500 g
UN Number UN1891
Beilstein 1209224
Sensitivity Light sensitive
Merck Index 14,3771
Solubility Information Miscible with ethanol,ether,chloroform and organic solvents.
Formula Weight 108.97
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Bromoethane