Learn More
Bromophenol Blue, for Electrophoresis, Fisher Chemical™
Product Code. p-7072889 Shop All Fisher Chemical Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. 10243420
Brand: Fisher Chemical B/P620/44
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Melting Point 279°C
CAS 115-39-9
Molecular Formula C19H10Br4O5S
MDL Number 5875
Formula Weight 669.96g/mol
Packaging Glass Jar
Color Red to Violet
Quantity 5 g
Physical Form Solid