missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brorin/VWC2 Antibody (08), DyLight 405 , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-05797V
This item is not returnable.
View return policy
Description
Brorin/VWC2 Monoclonal antibody specifically detects Brorin/VWC2 in Human samples. It is validated for ELISA, ELISA
Specifications
| Brorin/VWC2 | |
| Monoclonal | |
| DyLight 405 | |
| Brain-Specific Chordin-Like, Brain-Specific Chordin-Like Protein, brorin, PSST739, UNQ739, Von Willebrand Factor C Domain Containing 2, Von Willebrand Factor C Domain-Containing Protein 2 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Brorin/VWC2 (NP_940972.2; Met1-Met325). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 08 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 375567 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction