Learn More
Hirschmann™ Class A Amber Glass Volumetric Flasks with Poly Stopper
Product Code. 13177939
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
Standard Taper Joint:
10/19 NS Standard Ground
12/21 NS Standard Ground
14/23 NS Standard Ground
19/26 NS Standard Ground
24/29 NS Standard Ground
29/32 NS Standard Ground
7/16 NS Standard Ground
Tolerance:
±0.02 mL
±0.025 mL
±0.03 mL
±0.04 mL
±0.06 mL
±0.08 mL
±0.1 mL
±0.15 mL
±0.25 mL
±0.4 mL
±0.6 mL
Unit Size:
Each
Pack of 2
Product Code. 13177939
Brand: Hirschmann™ 2640161
Allows for precise, safe liquid measurement and handling of light-sensitive materials in the laboratory. Hirschmann Instruments™ Class A Amber Glass Volumetric Flasks with Poly Stopper are manufactured from durable DURAN™ glass tubing.
  • With NS ground and polyethylene stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch identification
Specifications
Certifications/Compliance DIN EN ISO 1042
Closure Material Polyethylene
Color Brown
Capacity (Metric) 10 mL
Material Borosilicate Glass, Duran Glass
Height (Metric) 90 mm
Limit of Error ±0.04 mL
Standard Taper Joint 10/19 NS Standard Ground
Tolerance ±0.04 mL
Graduated Yes
Class A