Learn More
Kautex™ Brown Low Density Polyethylene Wide Neck Bottles
Use brown-transparent Kautex™ Brown Low Density Polyethylene Wide Neck Bottles to limit the exposure to light and lessen possible tainting of bottle contents through light transmission.
Brand: Kautex™ 2000784585
Additional Details : Weight : 7.02000kg
Description
- Brown color protects light-sensitive materials against UV-light
- Wide neck opening means bottles are easy to fill and clean
- Round
- With closure
Specifications
LDPE | |
120 mm | |
Screw Cap | |
250 mL | |
30.8 mm | |
LDPE | |
260 | |
10 Pk. | |
No |
Wide Neck Bottle | |
64 mm | |
40 mm | |
Brown | |
Round | |
30.8 mm | |
Screw Closure LDPE | |
Yes |