Learn More
Invitrogen™ BRPF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5120519
Description
Positive test controls include: Mouse testis. Immunogen sequence: MGVDFDVKTF CHNLRATKPP YECPVETCRK VYKSYSGIEY HLYHYDHDNP PPPQQTPLRK HKKKGRQSRP ANKQSPSPSE VSQSPGREVM SYAQAQRMVE VDLHGRVHRI SIFDNLDVVS EDEEAPEEAP ENGSNKENTE TPAATPKSGK HKNKEKRKDS NHHHHHNVSA STTPKLPEVV YRELEQDTPD APPRPTSYYR.
BRPF1 (peregrin) is a component of the MOZ/MORF complex. This complex has histone H3 acetyltransferase activity. BRPF1 preferentially mediates histone H3-K23 acetylation and positively regulates the transcription of RUNX1 and RUNX2. Mutations affecting the gene can result in intellectual developmental disorder with dysmorphic facies and ptosis.
Specifications
| BRPF1 | |
| Polyclonal | |
| Unconjugated | |
| BRPF1 | |
| 4833438B11Rik; 4930540D11Rik; BR140; bromodomain and PHD finger containing 1; bromodomain and PHD finger containing, 1; bromodomain and PHD finger-containing protein 1; bromodomain-containing protein, 140kD; Brpf1; Brpf2; Peregrin; Protein Br140 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 679713, 7862, 78783 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.74 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| B2RRD7, P55201 | |
| BRPF1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human BRPF1 (NP_001003694). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.