missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRPF3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92324-0.02ml
This item is not returnable.
View return policy
Description
BRPF3 Polyclonal antibody specifically detects BRPF3 in Rat samples. It is validated for Western Blot
Specifications
| BRPF3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| bromodomain and PHD finger containing, 3, bromodomain and PHD finger-containing protein 3, KIAA1286 | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human BRPF3 (NP_056510.2). HRISIYDPLKIITEDELTAQDITECNSNKENSEQPQFPGKSKKPSSKGKKKESCSKHASGTSFHLPQPSFRMVDSGIQPEAPPLPAAYYRYIEKPPEDLDA | |
| 0.02 mL | |
| Cell Biology | |
| 27154 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction