missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRSK1 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98618AF647
This item is not returnable.
View return policy
Description
BRSK1 Polyclonal specifically detects BRSK1 in Human samples. It is validated for Western Blot.
Specifications
| BRSK1 | |
| Polyclonal | |
| Western Blot | |
| BR serine/threonine kinase 1, EC 2.7.11, EC 2.7.11.1, FLJ43009, KIAA1811BR serine/threonine-protein kinase 1, protein kinase SAD1A, SAD1, SAD1 kinase, Serine/threonine-protein kinase SAD-B | |
| Produced in rabbits immunized with E. coli-derived Human BRSK1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 84446 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction