missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRSK2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77281-0.025mg
This item is not returnable.
View return policy
Description
BRSK2 Polyclonal specifically detects BRSK2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| BRSK2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| BR serine/threonine kinase 2, BR serine/threonine-protein kinase 2, C11orf7, chromsosome 11 open reading frame 7, EC 2.7.11, EC 2.7.11.1, FLJ41362, PEN11BSAD1, protein kinase SAD1B, serine/threonine kinase 29, serine/threonine kinase SAD-A, Serine/threonine-protein kinase 29, Serine/threonine-protein kinase SAD-A, STK29 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence 20 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL | |
| Q8IWQ3 | |
| BRSK2 | |
| Antibody was raised against a 23 amino acid synthetic peptide from near the carboxy terminus of human BRSK2. The immunogen is located within amino acids 630 - 680 of BRSK2. Amino Acid Squence: DPPAAQHLSDTTNNCMEMMTGRLS | |
| 0.025 mg | |
| Immune System Diseases, Immunology, Protein Kinase | |
| 9024 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction