missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brucella Abortus Antibody (BrG11) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-73111JF669
This item is not returnable.
View return policy
Description
Brucella Abortus Monoclonal antibody specifically detects Brucella Abortus in Bacteria samples. It is validated for ELISA,Western Blot
Specifications
| Brucella Abortus | |
| Monoclonal | |
| Janelia Fluor 669 | |
| B.abortus | |
| This is reacting with intact Brucella-cells. | |
| 0.2 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG2a |
| ELISA, Western Blot | |
| BrG11 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Bacteria | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction