missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BRUNOL4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15796071 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15796071 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15796071 Supplier Invitrogen™ Supplier No. PA558196

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MYIKMATLAN GQADNASLST NGLGSSPGSA GHMNGLSHSP GNPSTIPMKD H Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

Members of the CELF/BRUNOL protein family contain two N-terminal RNA recognition motif (RRM) domains, one C-terminal RRM domain, and a divergent segment of 160-230 aa between the second and third RRM domains. Members of this protein family regulate pre-mRNA alternative splicing and may also be involved in mRNA editing, and translation. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BRUNOL4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene CELF4
Gene Accession No. Q9BZC1
Gene Alias A230070D14Rik; Brul4; Bruno -like 4; brunol4; BRUNOL-4; bruno-like 4, RNA binding protein; bruno-like protein 4; C130060B05Rik; celf4; CELF-4; CUGBP; CUG-BP and ETR-3 like factor 4; CUG-BP- and ETR-3-like factor 4; CUGBP Elav-like family member 4; CUGBP, Elav-like family member 4; LYST-interacting protein LIP9; RNA-binding protein BRUNOL4; RNA-binding protein BRUNOL-4; zgc:92761
Gene Symbols CELF4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human BRUNOL4. Recombinant protein control fragment (Product #RP-96548).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56853
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.