Learn More
Product Code | Brand | Size | Product Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Product Type | Price | Quantity & Availability | ||||
10068250
|
Vereinigte Pinselfabriken™
620-0,7 |
0.75 in. | Type 620 Brush |
€ 5.38
Each |
Estimated Shipment: 03-05-2024
Log in to see stock. |
||||
10788911
|
Vereinigte Pinselfabriken™
620-1,0 |
25.4 mm | Type 620 Brush |
POA
Each |
N/A | ||||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
Specifications
1 Ea. |