missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BSDC1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18457B
This item is not returnable.
View return policy
Description
BSDC1 Polyclonal antibody specifically detects BSDC1 in Human samples. It is validated for Western Blot
Specifications
| BSDC1 | |
| Polyclonal | |
| Western Blot | |
| BSD domain containing 1, UNQ2494/PRO5781 | |
| Recognizes region between residues Between Between 380 and 430.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55108 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction