missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BSDC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542193
This item is not returnable.
View return policy
Description
Peptide sequence: VISDTFAPSP DKTIDCDVIT LMGTPSGTAE PYDGTKARLY SLQSDPATYC Sequence homology: Cow: 100%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
BSDC1 (BSD Domain Containing 1) is a Protein Coding gene. An important paralog of this gene is SYAP1. [GeneCards].
Specifications
| BSDC1 | |
| Polyclonal | |
| Unconjugated | |
| BSDC1 | |
| 1110063F24Rik; AW011758; BSD domain containing 1; BSD domain-containing protein 1; BSDC1; DKFZp686B09139; FLJ10276; hypothetical protein LOC505545; RGD1311622; RP4-811H24.7; UNQ2494/PRO5781 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 55108 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9NW68 | |
| BSDC1 | |
| Synthetic peptide directed towards the N-terminal of human BSDC1 (aa 84-133). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction