missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BSSP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5103666
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PRSS22.
This gene encodes a member of the trypsin family of serine proteases. The enzyme is expressed in the airways in a developmentally regulated manner. The gene is part of a cluster of serine protease genes on chromosome 16.
Specifications
| BSSP4 | |
| Polyclonal | |
| Unconjugated | |
| PRSS22 | |
| 4733401N09Rik; brain-specific serine protease 4; BSP-2; BSSP4; BSSP-4; hBSSP-4; MGC9599; prosemin; protease, serine 22; protease, serine S1 family member 22; protease, serine, 22; PRSS22; Prss26; Serine protease 22; serine protease 26; SP001LA; Tryptase epsilon; UNQ302/PRO343 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 64063, 70835 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9ER10, Q9GZN4 | |
| PRSS22 | |
| A synthesized peptide derived from human PRSS22(Accession Q9GZN4), corresponding to amino acid residues L278-S317. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction