missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BST2 Antibody (2E2), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-29621F
This item is not returnable.
View return policy
Description
BST2 Monoclonal specifically detects BST2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| BST2 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| BST2 | |
| Full length recombinant human BST2 protein [NCBI NP_004326] | |
| 0.1 mL | |
| Signal Transduction | |
| 684 | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 200 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| bone marrow stromal antigen 2, bone marrow stromal cell antigen 2, BST-2, CD317, CD317 antigen, HM1.24 antigen, NPC-A-7, PDCA-1, TETHERIN | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction