missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BST2 Antibody (44E9R), PE/Cy7, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB8660PECY7
This item is not returnable.
View return policy
Description
BST2 Monoclonal antibody specifically detects BST2 in Mouse samples. It is validated for Flow Cytometry
Specifications
| BST2 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 44E9R | |
| Flow Cytometry | |
| bone marrow stromal antigen 2, bone marrow stromal cell antigen 2, BST-2, CD317, CD317 antigen, HM1.24 antigen, NPC-A-7, PDCA-1, TETHERIN | |
| Recombinant extracellular domain of CD317, | |
| 0.1 mL | |
| Signal Transduction | |
| 684 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction