missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BST2 Antibody (4F6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-29622CL1
This item is not returnable.
View return policy
Description
BST2 Monoclonal antibody specifically detects BST2 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Flow (Cell Surface),CyTOF
Specifications
| BST2 | |
| Monoclonal | |
| CoraFluor 1 | |
| bone marrow stromal antigen 2, bone marrow stromal cell antigen 2, BST-2, CD317, CD317 antigen, HM1.24 antigen, NPC-A-7, PDCA-1, TETHERIN | |
| Full length recombinant human BST2 protein [NCBI NP_004326]. | |
| 0.1 mL | |
| Signal Transduction | |
| 684 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Flow Cytometry (Cell Surface) | |
| 4F6 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction