Learn More
BST2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13560-1-AP-150UL
 View more versions of this product
Product Code. 16871834
  • € 410.00 / 150µL
Description
Bone marrow stromal cells are involved in the growth and development of B-cells. The specific function of the protein encoded by the bone marrow stromal cell antigen 2 is undetermined; however, this protein may play a role in pre-B-cell growth and in rheumatoid arthritis.
Specifications
Specifications
BST2
Polyclonal
Unconjugated
BST2
Bone marrow stromal antigen 2, BST 2, BST2, CD317, HM1.24 antigen, Tetherin
Rabbit
Antigen Affinity Chromatography
RUO
684
-20°C
Liquid
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q10589
BST2
BST2 Fusion Protein Ag4430
150 μL