Learn More
BST2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13560-1-AP-150UL
This item is not returnable.
View return policy
Description
Bone marrow stromal cells are involved in the growth and development of B-cells. The specific function of the protein encoded by the bone marrow stromal cell antigen 2 is undetermined; however, this protein may play a role in pre-B-cell growth and in rheumatoid arthritis.Specifications
| BST2 | |
| Polyclonal | |
| Unconjugated | |
| BST2 | |
| Bone marrow stromal antigen 2, BST 2, BST2, CD317, HM1.24 antigen, Tetherin | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 684 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q10589 | |
| BST2 | |
| BST2 Fusion Protein Ag4430 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |