missing translation for 'onlineSavingsMsg'
Learn More

BTAF1 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NB100-57501JF646

Product Code. 18721313

  • € 633.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

BTAF1 Polyclonal antibody specifically detects BTAF1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BTAF1
Polyclonal
Western Blot
ATP-dependent helicase BTAF1, BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170kDa (Mot1homolog, S. cerevisiae), B-TFIID transcription factor-associated 170 kDa subunit, EC 3.6.1, EC 3.6.4.-, MOT1MGC138406, TAF(II)170KIAA0940, TAF-172, TAF172BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170 kD (Mot1homolog, S. cerevisiae), TAFII170TATA-binding protein-associated factor 172, TBP-associated factor 172
The immunogen recognized by this antibody maps to a region between residue 75 and 125 of human RNA polymerase II, B-TFIID transcription factor-associated, 170kDa using the numbering given in entry NP_003963.1 (GeneID 9044).
0.1 mL
Primary
Human
Purified
Western Blot
Janelia Fluor 646
50mM Sodium Borate
Rabbit
Immunogen affinity purified
RUO
9044
Store at 4C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.