missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTBD15 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 462.00
Specifications
| Antigen | BTBD15 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
BTBD15 Polyclonal antibody specifically detects BTBD15 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| BTBD15 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| BTB (POZ) domain containing 15, BTB/POZ domain-containing protein 15, BTBD15, HSPC063, MGC57431, MGC60348, MGC88058, zinc finger and BTB domain containing 44, zinc finger and BTB domain-containing protein 44, Zinc finger protein 851, ZNF851MGC26123 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ZBTB44 (NP_001288027.1). MGVKTFTHSSSSHSQEMLGKLNMLRNDGHFCDITIRVQDKIFRAHKVVLAACSDFFRTKLVGQAEDENKNVLDLHHVTVTGFIPLLEYAYTATLSINTENIIDVLAAASYMQMFSVASTCSEFMKSSILWNTPNSQPEKGLDAGQENNSNCNFTSRDGSISPVSSECSVVERTIPVCRESRRKRKSYIVMSPESPVKCGTQTSSPQVLNSSASYSENRNQPVDSSLAFPWTFPFGIDRRIQPEKVKQAENTRTLELPGPSETGRRMADYVTCESTKTTLPLGTEEDVRVKVERLSDEEVH | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 50% glycerol, pH7.3. | |
| 29068 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title