missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BTBD16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578632
This item is not returnable.
View return policy
Description
Positive Control: NT2D1 Predicted Reactivity: Chimpanzee (95%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
BTBD16 encodes a protein that contains a BTB/POZ domain. This domain mediates protein-protein interactions. A mutation in this gene may be associated with bipolar disorder.
Specifications
| BTBD16 | |
| Polyclonal | |
| Unconjugated | |
| BTBD16 | |
| BTB (POZ) domain containing 16; BTB domain containing 16; BTB/POZ domain-containing protein 16; Btbd16; C10orf87; E330040A16Rik; FLJ25359; MGC129864; MGC129865 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 118663 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q32M84 | |
| BTBD16 | |
| Recombinant protein encompassing a sequence within the center region of human BTBD16. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction