missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTBD3 Antibody (OTI1A10), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71699IR
This item is not returnable.
View return policy
Description
BTBD3 Monoclonal specifically detects BTBD3 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| BTBD3 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BTBD3 | |
| Full length recombinant protein of human BTBD3 (NP_852108) produced in E.coli. | |
| 0.1 mL | |
| 22903.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI1A10 | |
| Western Blot | |
| BTB (POZ) domain containing 3, BTB/POZ domain-containing protein 3, KIAA0952dJ742J24.1, MGC130038, MGC130039 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction