missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BTBD6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA549871
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total BTBD6 protein.
Adapter protein for the CUL3 E3 ubiquitin-protein ligase complex. Involved in late neuronal development and muscle formation.
Specifications
| BTBD6 | |
| Polyclonal | |
| Unconjugated | |
| BTBD6 | |
| BDPL; BTB (POZ) domain containing 6; BTB domain containing 6; BTB domain protein BDPL; BTB/POZ domain-containing protein 6; Btbd6; glucocorticoid receptor AF-1 coactivator-1; lens BTB domain protein; LOW QUALITY PROTEIN: BTB/POZ domain-containing protein 6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 399566, 90135 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8K2J9, Q96KE9 | |
| BTBD6 | |
| Synthesized peptide derived from interf human BTBD6 (aa 91-140). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction