missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTBD9 Antibody (BTBD9/7501) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-24035JF669
This item is not returnable.
View return policy
Description
BTBD9 Monoclonal antibody specifically detects BTBD9 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| BTBD9 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| BTB (POZ) domain containing 9, BTB/POZ domain-containing protein 9, FLJ32945, KIAA1880dJ322I12.1, MGC120517, MGC120519, MGC120520 | |
| Recombinant fragment (around aa200-400) of human BTBD9 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| BTBD9/7501 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 114781 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction