missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTF Antibody (M33-P5B11), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50118DL594
This item is not returnable.
View return policy
Description
BTF Monoclonal antibody specifically detects BTF in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| BTF | |
| Monoclonal | |
| DyLight 594 | |
| bcl-2-associated transcription factor 1, BCL2-associated transcription factor 1, Btf, BTFKIAA0164bK211L9.1 | |
| Peptide Sequence - KYQGDGIVED (amino acids 900 - 909) | |
| 0.1 mL | |
| Apoptosis, DNA Repair | |
| 9774 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| M33-P5B11 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction