missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTF3 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-28700B
This item is not returnable.
View return policy
Description
BTF3 Polyclonal antibody specifically detects BTF3 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| BTF3 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry | |
| basic transcription factor 3, BETA-NAC, BTF3a, BTF3b, NACB, nascent-polypeptide-associated complex beta polypeptide, RNA polymerase B transcription factor 3, transcription factor BTF3 | |
| The immunogen recognized by this antibody maps to a region between residue 35 and 70 of basic transcription factor 3 using the numbering given in entry NP_001032726.1 (GeneID 689). | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 689 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction