missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTG1 Antibody (OTI7H6), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70286AF488
This item is not returnable.
View return policy
Description
BTG1 Monoclonal specifically detects BTG1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| BTG1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BTG1 | |
| Full length recombinant protein of human BTG1 (NP_001722) produced in E.coli. | |
| 0.1 mL | |
| 694.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI7H6 | |
| Immunohistochemistry | |
| B-cell translocation gene 1 protein, B-cell translocation gene 1, proliferative, protein BTG1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction