Learn More
BTG2 Monoclonal Antibody (OTI9D3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808153S
Encompass_Preferred
 View more versions of this product
Product Code. 17519103
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
BTG2 Monoclonal antibody specifically detects BTG2 in Human samples. It is validated for Western Blot
Specifications
Specifications
BTG2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P78543
Mouse
Affinity chromatography
RUO
7832
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI9D3
Unconjugated
BTG2
AA959598; Agl; An; an-1; APRO1; B cell translocation gene 2, anti-proliferative; B-cell translocation gene 2; B-cell translocation gene 2, anti-proliferative; BTG anti-proliferation factor 2; BTG family member 2; BTG family, member 2; BTG2; cell surface alloantigen; Early induced gene B-cell translocation gene 2; nerve growth factor-inducible anti-proliferative; NGF-inducible anti-proliferative protein PC3; NGF-inducible anti-proliferative putative secreted protein; NGF-inducible protein TIS21; Pc3; pheochromacytoma cell-3; protein BTG2; Tis21
Full length human recombinant protein of human BTG2 produced in E.coli.
30 μL