Learn More
Invitrogen™ BTG4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113416
This item is not returnable.
View return policy
Description
BTG4 Polyclonal Antibody for Western Blot, IHC (P), ELISA
Shows marked antiproliferative activity, being able to induce G(1) arrest.
Specifications
| BTG4 | |
| Polyclonal | |
| Unconjugated | |
| Btg4 | |
| APRO3; B cell translocation gene 4; B-cell translocation gene 4; BTG anti-proliferation factor 4; BTG family member 4; BTG4; C86116; PC3B; Protein BTG4; protein PC3b; SCIR-27 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 54766, 56057 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2.63 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| O70552, Q9NY30 | |
| Btg4 | |
| Recombinant Human Protein BTG4 protein (1-206AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |