missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTLA/CD272 Antibody (753131), DyLight 680, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB7600Y
This item is not returnable.
View return policy
Description
BTLA/CD272 Monoclonal antibody specifically detects BTLA/CD272 in Mouse samples. It is validated for Western Blot
Specifications
| BTLA/CD272 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| 753131 | |
| Western Blot | |
| B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 | |
| Mouse myeloma cell line NS0-derived recombinant mouse BTLA, Met1-Pro176, Accession # Q7TSA3 | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 151888 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction