missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTLA/CD272 Antibody (922701), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB33541PCP
This item is not returnable.
View return policy
Description
BTLA/CD272 Monoclonal specifically detects BTLA/CD272 in Human samples. It is validated for Flow Cytometry.
Specifications
| BTLA/CD272 | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| BTLA | |
| rhBTLA aa25-150 | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 922701 | |
| Flow Cytometry | |
| B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 151888 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction