missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTLA/CD272 Antibody (HMBT-6B2), CoraFluor™ 1, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-81117CL1
This item is not returnable.
View return policy
Description
BTLA/CD272 Monoclonal antibody specifically detects BTLA/CD272 in Mouse samples. It is validated for Flow Cytometry,Immunoprecipitation,Western Blot,Functional Assay,Immunoblot,Neutralization
Specifications
| BTLA/CD272 | |
| Monoclonal | |
| CoraFluor 1 | |
| B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 | |
| This antibody was raised by immunising Armenian hamsters with mouse BTLA/CD272:Fc fusion protein. | |
| 0.1 mL | |
| Cell Cycle and Replication, Phospho Specific | |
| 151888 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG κ |
| Flow Cytometry, Immunoprecipitation, Western Blot, Functional Assay, Immunoblot | |
| HMBT-6B2 | |
| PBS | |
| Armenian Hamster | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction